Please login to your online account to display your discounted pricing

Alfa Aesar™ Guanidineacetic acid, 99+%

$55.14 - $55.14

Chemical Identifiers

Chemical Name or Material Guanidineacetic acid
CAS 352-97-6
Molecular Formula C3H7N3O2
Formula Weight 117.11
MDL Number MFCD00004278
Beilstein 1759179
Merck Index 14,4495
Synonym N-Amidinoglycine; Glycocyamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1628114

  SDS
alfa aesar™
L1628114
25g Each for $55.14
Description & Specifications

Specifications

Formula Weight 117.11
CAS 352-97-6
Health Hazard 1 Warning
Assay ≥99%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Merck Index 14,4495
Beilstein 1759179
MDL Number MFCD00004278
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C3H7N3O2
Synonym N-Amidinoglycine; Glycocyamine
Melting Point ≈280°C
Quantity 25g
Chemical Name or Material Guanidineacetic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.