Please login to your online account to display your discounted pricing

Guanidine Sulfate 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  411101000

 View more versions of this product

Catalog No. AC411101000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 216.22
Melting Point >245°C
Quantity 100g
Color White
EINECS Number 209-826-8
TSCA TSCA
Appearance of Solution (50% soln. in water) clear
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Packaging Glass bottle
Sulfated Ash 0.1% max.
Water 0.5% max. (K.F.)
Chemical Name or Material Guanidine sulfate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C H5 N3 . 0.5 H2 S O4
Linear Formula [NH2C(NH)NH2]2·H2SO4
MDL Number MFCD00013115
CAS 594-14-9

Specifications

Chemical Name or Material Guanidine sulfate
Name Note 99%
CAS 594-14-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C H5 N3 . 0.5 H2 S O4
Linear Formula [NH2C(NH)NH2]2·H2SO4
Formula Weight 216.22
MDL Number MFCD00013115