Please login to your online account to display your discounted pricing

Guanidine Sulfate 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Guanidine sulfate
Name Note 99%
CAS 594-14-9
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C H5 N3 . 0.5 H2 S O4
Linear Formula [NH2C(NH)NH2]2·H2SO4
Formula Weight 216.22
MDL Number MFCD00013115
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC411101000

  SDS
fisher scientific acros
411101000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC411105000

  SDS
fisher scientific acros
411105000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 216.22
Melting Point >245°C
Quantity 100g
Color White
EINECS Number 209-826-8
TSCA TSCA
Appearance of Solution (50% soln. in water) clear
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Packaging Glass bottle
Sulfated Ash 0.1% max.
Water 0.5% max. (K.F.)
Chemical Name or Material Guanidine sulfate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C H5 N3 . 0.5 H2 S O4
Linear Formula [NH2C(NH)NH2]2·H2SO4
MDL Number MFCD00013115
CAS 594-14-9