Please login to your online account to display your discounted pricing

Guanidine Hydrochloride GenAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Guanidine Hydrochloride
CAS 50-01-1
Molecular Formula NH2C(:NH)NH2·HCl
Formula Weight 95.53
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK771687

 
avantor performance
771687
Poly Pail 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 95.53
Quantity 10kg
Molecular Formula NH2C(:NH)NH2·HCl
Chemical Name or Material Guanidine Hydrochloride
Packaging Poly Pail
CAS 50-01-1