missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™

Catalog No. BP178500
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP178500 500 g
BP1781 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP178500 Supplier Fisher BioReagents Supplier No. BP178 500

For isolation of RNA from cell extracts | CAS: 50-01-1 | CH6ClN3

For isolation of RNA from cell extracts

Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.

Specifications

Chemical Name or Material Guanidine Hydrochloride
CAS 50-01-1
Assay Percent Range ≥99.5%
Synonym guanidine hydrochloride
Quantity 500 g
Merck Index 15, 4597
Melting Point 178°C
Molecular Formula CH6ClN3
Formula Weight 95.53
Physical Form Solid
Color White
Purity ≥99.5%
Show More Show Less