missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™

For isolation of RNA from cell extracts

$135.23 - $234.38

Specifications

Chemical Name or Material Guanidine Hydrochloride
CAS 50-01-1
Assay Percent Range ≥99.5%
Synonym guanidine hydrochloride
Merck Index 15, 4597
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP178-500
View Documents Promotion Details
Fisher BioReagents
BP178500
500 g
Each for $135.23
Case of 6 Each for $811.38
 
BP178-1 Edge
View Documents Promotion Details
Fisher BioReagents
BP1781
1 kg
Each for $234.38
Case of 6 Each for $1,406.28
Edge
 
Description

Description

For isolation of RNA from cell extracts

Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.
Specifications

Specifications

Guanidine Hydrochloride
≥99.5%
15, 4597
CH6ClN3
Solid
≥99.5%
50-01-1
guanidine hydrochloride
178°C
95.53
White
SDS