Please login to your online account to display your discounted pricing

Guaiac Tincture, 1:60 in Alcohol, for Blood Detection, LabChem™

$140.77 - $140.77

Chemical Identifiers

Chemical Name or Material Guaiac Tincture
CAS 64-17-5
CAS Max % 84.1
CAS 7732-18-5
CAS Max % 4.92
CAS 67-63-0
CAS Max % 4.67
CAS 67-56-1
CAS Max % 4.67
CAS 9000-29-7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC149151

 
lab chem
LC149151
500mL Each for $140.77
Description & Specifications

Specifications

CAS 64-17-5
Name Note 1:60 in Alcohol, for Blood Detection
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 67-63-0
Health Hazard 1 Danger
Identification Passes Test
Chemical Name or Material Guaiac Tincture
CAS Max % 4.67
CAS Max % 1.64
Solubility Soluble in water
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Physical Form Liquid
Storage Information Room Temperature
CAS 67-56-1
CAS Max % 4.67
CAS 9000-29-7
CAS 7732-18-5
Color Clear, dark brown
Quantity 500mL
CAS Max % 4.92
CAS Max % 84.1
Packaging Poly Bottle
UN Number UN1170
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).