missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GTF2I, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002969B01P
Description
Sequence: MAQVAMSTLPVEDEESSESRMVVTFLMSALESMCKELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGISFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGPSETDDVDEKQPLSKPLQGSHHSSEGNEGTEMEVPAEDDDYSPPSKRPKANELPQPPVPEPANAGKRKVREFNFEKWNARITDLRKQVEELFERKYAQAIKAKGPVTIPYPLFQSHVEDLYVEGLPEGIPFRRPSTYGIPRLERILLAKERIRFVIKKHELLNSTREDLQLDKPASGVKEEWYARITKLRKMVDQLFCKKFAEALGSTEAKAVPYQKFEAHPNDLYVEGLPENIPFRSPSWYGIPRLEKIIQVGNRIKFVIKRPELLTHSTTEVTQPRTNTPVKEDWNVRITKLRKQVEEIFNLKFAQALGLTEAVKVPYPVFESNPEFLYVEGLPEGIPFRSPTWFGIPRLERIVRGSNKIKFVVKKPELVISYLPPGMASKINTKALQSPKRPRSPGSNSKVPEIEVTVEGPNNNNPQTSAVRTPTQTNGSNVPFKPRGREFSFEAWNAKITDLKQKVENLFNEKCGEALGLKQAVKVPFALFESFPEDFYVEGLPEGVPFRRPSTFGIPRLEKILRNKAKIKFIIKKPEMFETAIKESTSSKSPPRKINSSPNVNTTASGVEDLNIIQVTIPDDDNERLSKVEKARQLREQVNDLFSRKFGEAIGMGFPVKVPYRKITINRGCVVVDGMPPGVSFKAPSYLEISSMRRILDSAEFIKFTVIRPFPGLVVNNQLVDQSESEGPVIQESAEPSQLEVPATEEIKETDGSSQIKQEPDPTWSpecifications
GTF2I | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC099907.1 | |
GTF2I | |
GTF2I (AAH99907.1, 1 a.a. ∼ 976 a.a) full-length human protein. | |
RUO | |
2969 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunofluorescence,Western Blot | |
Unconjugated | |
general transcription factor II, i | |
BAP-135/BAP135/BTKAP1/DIWS/FLJ38776/FLJ56355/IB291/SPIN/TFII-I/WBS/WBSCR6 | |
Mouse | |
50 μg | |
Primary | |
Human,Mouse | |
Antibody |