Please login to your online account to display your discounted pricing

GT 2016, Tocris

Selective H3 antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material GT 2016
Assay Percent Range >98%
Molecular Formula C19H31N3O
Formula Weight 317.47
Synonym 5-Cyclohexyl-1-[4-(1H-imidazol-5-yl)-1-piperidinyl]-1-pentanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

241910

 
r & d systems
RDS241910
10mg Each for $214.12

241950

 
r & d systems
RDS241950
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 317.47
Quantity 10mg
Chemical Name or Material GT 2016
Assay Percent Range >98%
Molecular Formula C19H31N3O
Synonym 5-Cyclohexyl-1-[4-(1H-imidazol-5-yl)-1-piperidinyl]-1-pentanone
CAS 152241-24-2
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C