missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and affinity purification applications with Thermo Scientific™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody.

$244.18 - $1,194.02

Specifications

Antigen GST (Epitope Tag) Ab-1
Clone GST01 (794)
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS707P Thermo Scientific™ Lab Vision™
MS707P
200μg/mL 1mL Each for $1,132.96
MS707P0 Thermo Scientific™ Lab Vision™
MS707P0
200μg/mL 100μL Each for $244.18
MS707P1 Thermo Scientific™ Lab Vision™
MS707P1
200μg/mL 500μL Each for $703.41
MS707P1ABX Thermo Scientific™ Lab Vision™
MS707P1ABX
1mg/mL 100μL Each for $797.16
MS707PABX Thermo Scientific™ Lab Vision™
MS707PABX
1mg/mL 200μL Each for $1,194.02
Specifications

Description

Ensure accurate, reproducible results in western blotting and affinity purification applications with Thermo Scientific™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: GST01; same as 794

Isotype: IgG1

Species Reactivity: Proteins from all species that are tagged with GST

Immunogen: Purified recombinant GST protein

Cellular Localization: Depends upon the localization of the parent protein tagged with GST

Recommended for:

  • Western Blotting
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

GST (Epitope Tag) Ab-1
Western Blot
Unlabeled
Murine
Enzyme
Purified recombinant GST protein
GST01 (794)
Monoclonal
Concentrated
RUO
Primary
IgG1