Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting and affinity purification applications with Thermo Scientific™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS707P1ABX

 View more versions of this product

Catalog No. MS707P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Clone GST01 (794)
Format Concentrated
Applications Western Blotting
Quantity 100µL
Cross Reactivity All
Antigen GST (Epitope Tag) Ab-1
Immunogen Purified recombinant GST protein
Isotype IgG1
Regulatory Status RUO
Conjugate Unlabeled
Research Discipline Enzyme
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Ensure accurate, reproducible results in western blotting and affinity purification applications with Thermo Scientific™ GST (Epitope Tag) Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: GST01; same as 794

Isotype: IgG1

Species Reactivity: Proteins from all species that are tagged with GST

Immunogen: Purified recombinant GST protein

Cellular Localization: Depends upon the localization of the parent protein tagged with GST

Recommended for:

  • Western Blotting
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO