Please login to your online account to display your discounted pricing

GSK2578215A, Tocris™

Potent, selective LRRK2 inhibitor; brain penetrant

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material GSK2578215A
Assay Percent Range >99%
Molecular Formula C24H18FN3O2
Formula Weight 399.42
Synonym 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

462910

 
r & d systems
RDS462910
10mg Each for $284.81

462950

 
r & d systems
RDS462950
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 399.42
Quantity 10mg
Chemical Name or Material GSK2578215A
Assay Percent Range >99%
Molecular Formula C24H18FN3O2
CAS 1285515-21-0
Synonym 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature