Please login to your online account to display your discounted pricing

GSK J4, Tocris™

Histone demethylase JMJD3/UTX inhibitor; cell permeable

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material GSK J4
Assay Percent Range >99%
Molecular Formula C24H27N5O2
Formula Weight 417.5
Synonym N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

459450

 
r & d systems
459450
50mg Each for $1,073.10

459410

 
r & d systems
459410
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 417.5
Quantity 50mg
Chemical Name or Material GSK J4
Assay Percent Range >99%
Molecular Formula C24H27N5O2
CAS 1373423-53-0
Synonym N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine ethyl ester
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature