Please login to your online account to display your discounted pricing

GSK J1, Tocris™

Potent histone demethylase JMJD3/UTX inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material GSK J1
Assay Percent Range >99%
Molecular Formula C22H23N5O2
Formula Weight 389.45
Synonym N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

459310

 
r & d systems
459310
10mg Each for $257.85

459350

 
r & d systems
459350
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 389.45
Quantity 10mg
Chemical Name or Material GSK J1
Assay Percent Range >99%
Molecular Formula C22H23N5O2
Synonym N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine
Recommended Storage Desiccate at Room Temperature
CAS 1373422-53-7
Safety and Handling

Desiccate at Room Temperature