Please login to your online account to display your discounted pricing

GSK 9027, Tocris™

Glucocorticoid receptor agonist

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material GSK 9027
Assay Percent Range >98%
Molecular Formula C27H19F4N3O2S
Formula Weight 525.52
Synonym N-[4-[1-(4-Fluorophenyl)-1H-indazol-5-yl-3-(trifluoromethyl)phenyl]benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

411650

 
r & d systems
RDS411650
50mg Each for $1,116.26

411610

 
r & d systems
RDS411610
10mg Each for $265.15
Description & Specifications

Specifications

Chemical Name or Material GSK 9027
Quantity 50mg
CAS 1229096-88-1
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C27H19F4N3O2S
Formula Weight 525.52
Synonym N-[4-[1-(4-Fluorophenyl)-1H-indazol-5-yl-3-(trifluoromethyl)phenyl]benzenesulfonamide
Safety and Handling

Store at Room Temperature