Please login to your online account to display your discounted pricing

GSK 789472 hydrochloride, Tocris™

D3 antagonist; Also D2 partial agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material GSK 789472 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H21N3O·HCl
Formula Weight 295.81
Synonym 4, 5-Dihydro-1-phenyl-3-(2-piperidinyl)methylimidazol-2(1H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394050

 
r & d systems
RDS394050
50mg Each for $862.95

394010

 
r & d systems
RDS394010
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 295.81
Quantity 50mg
Chemical Name or Material GSK 789472 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H21N3O·HCl
CAS 1257326-24-1
Synonym 4, 5-Dihydro-1-phenyl-3-(2-piperidinyl)methylimidazol-2(1H)-one
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature