Please login to your online account to display your discounted pricing

Alfa Aesar™ GSK-3beta Peptide Inhibitor

For cellular and molecular biology applications

Specifications

Product Type GSK-3beta Peptide Inhibitor
Molecular Weight 1197.23
Storage Requirements -30° to -10°C
Sequence Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H)-Pro-NH2
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66645EXD

 
alfa aesar™
J66645EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66645MCR

 
alfa aesar™
J66645MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type GSK-3beta Peptide Inhibitor
Molecular Weight 1197.23
Storage Requirements -30° to -10°C
Sequence Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H)-Pro-NH2
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC