Please login to your online account to display your discounted pricing

Alfa Aesar™ GSK-3beta Peptide Inhibitor, Cell Permeable

For cellular and molecular biology applications

Specifications

Product Type GSK-3beta Peptide Inhibitor, cell permeable
Molecular Weight 1464.64
Storage Requirements -30° to -10°C
Sequence N-Myristoyl-Gly-Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H)-Pro-NH2
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66054EXD

 
alfa aesar™
J66054EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66054MCR

 
alfa aesar™
J66054MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type GSK-3beta Peptide Inhibitor, cell permeable
Molecular Weight 1464.64
Storage Requirements -30° to -10°C
Sequence N-Myristoyl-Gly-Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H)-Pro-NH2
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC