Please login to your online account to display your discounted pricing

GSK 3787, Tocris™

Potent and selective PPARδ antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material GSK 3787
Assay Percent Range >99%
Molecular Formula C15H12ClF3N2O3S
Formula Weight 392.78
Synonym 4-Chloro-N-[2-[[5-(trifluoromethyl)-2-pyridinyl]sulfonyl]ethyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

396110

 
r & d systems
RDS396110
10mg Each for $201.36

396150

 
r & d systems
RDS396150
50mg Each for $844.73
Description & Specifications

Specifications

Chemical Name or Material GSK 3787
Quantity 10mg
CAS 188591-46-0
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C15H12ClF3N2O3S
Formula Weight 392.78
Synonym 4-Chloro-N-[2-[[5-(trifluoromethyl)-2-pyridinyl]sulfonyl]ethyl]benzamide
Safety and Handling

Store at +4°C