Please login to your online account to display your discounted pricing

GSK 1059615, Tocris™

Potent PI 3-kinase inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material GSK 1059615
Assay Percent Range >98%
Molecular Formula C18H11N3O2S
Formula Weight 333.36
Synonym 5-[[4-(4-Pyridinyl)-6-quinolinyl]methylene]-2, 4-thiazolidenedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

402610

 
r & d systems
RDS402610
10mg Each for $257.85

402650

 
r & d systems
RDS402650
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 333.36
Quantity 10mg
Chemical Name or Material GSK 1059615
Assay Percent Range >98%
Molecular Formula C18H11N3O2S
CAS 958852-01-2
Synonym 5-[[4-(4-Pyridinyl)-6-quinolinyl]methylene]-2, 4-thiazolidenedione
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C