missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC GSC International Voltmeters

For numerous electrical experiments.

$38.50 - $41.00

Specifications

Description Full scale and fully enclosed
Product Type DC Voltmeter
For Use With (Application) For numerous electrical experiments.
View More Specs

Alert:

Not a toy. Product intended for ages 13 and up.



Products ${productFamilyLength}
Catalog Number Mfr. No. Voltage Price Quantity  
Catalog Number Mfr. No. Voltage Price Quantity  
S77811A GSC
4-130915
Range of 0-5 volts Each for $41.00
S77882A GSC
4-130918
0 to 3V, 0 to 10V, 0 to 15V Each for $38.50
Specifications

Description

  •  Accuracy of ±2.5%
  • Color-coded binding posts
  • Can be used with Banana cords
  • Range: 0 to 3V, 0 to 5V, 0 to 10V 0 to 15V, 0 to 300V
Description & Specifications

Specifications

Full scale and fully enclosed
DC Voltmeter
For numerous electrical experiments.