missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Pulleys

Useful tool for mechanical experiments.

Manufacturer:  GSC Go Science Crazy 41606215

 View more versions of this product

Catalog No. S40862A



Alert:

Not a toy. Product intended for ages 13 and up.


Description

Description

  • Quality product with aluminum housing

Double plastic pulley

  • 50mm dia. with an aluminum housing
Triple tandem pulley
  • With three wheels - dia. 27mm, 38mm, 50mm, Overall 7.5 in. L
Triple tandem pulley
  • With three wheels of various diameters with an aluminum housing
Quadruple pulley
  • With four wheels 50mm dia.

Specifications

Specifications

Double Pulley
Plastic
Two wheels
Documents