missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Penicillium Microscope Slide

Choose GSC prepared slides for your science classroom studies.

$6.35 - $6.80

Specifications

Grade 7 to College
Slide Type Pre-cleaned glass slides with smooth ground edges
Species Penicillium
Type Prepared Microscope Slides
Classification Fungi
View More Specs

Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S06375PS View Documents GSC Go Science Crazy
S06375PS
Each for $6.80
Pack of 10 Each for $68.00
 
S99232PS View Documents GSC Go Science Crazy
S99232PS
Each for $6.35
Pack of 10 Each for $63.50
 
Description

Description

  • Prepared slide for use with classroom science studies
Specifications

Specifications

7 to College
Penicillium
Fungi
Pre-cleaned glass slides with smooth ground edges
Prepared Microscope Slides