GSC Go Science Crazy Mammal Smooth Muscle Microscope Slide
Showing smooth fibers
$7.70 - $7.70
Specifications
Type Prepared Microscope Slides
Slide Type Pre-cleaned glass slides with smooth ground edges
For Use With (Application) Shows smooth fibers
View More Specs
Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S99066PS View Documents GSC Go Science Crazy
S99066PS
Each for $7.70
Pack of 10 Each for $77.00
Description
Description
  • Smooth muscle is found in walls of hollow organs such as intestinal tract, bladder and uterus 
  • They are neither striated in structure nor under voluntary control
  • Fibers have single nucleus and lack any striations
Specifications
Specifications
Prepared Microscope Slides
Pre-cleaned glass slides with smooth ground edges
Shows smooth fibers