missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Lilium Anther Microscope Slide

Choose GSC prepared slides for your science classroom studies.

$8.65 - $8.65

Specifications

Grade 7 to College
Slide Type Pre-cleaned glass slides with smooth ground edges
Species Lillium
Type Prepared Microscope Slides
Classification Botany
View More Specs

Products 1
Catalog Number Mfr. No. Specimen Type Price Quantity  
Catalog Number Mfr. No. Specimen Type Price Quantity  
S98969PS View Documents GSC Go Science Crazy
S98969PS
Each for $8.65
Pack of 10 Each for $86.50
 
Description

Description

  • Prepared slide for use with classroom science studies
Specifications

Specifications

7 to College
Lillium
Botany
Pre-cleaned glass slides with smooth ground edges
Prepared Microscope Slides
Cross section