missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Frog Striated Skeletal Muscle Microscope Slide

Shows striated muscle tissue

$6.28 - $6.28

Specifications

Type Prepared Microscope Slides
Species Frog
Classification Zoology
Grade 7 to College
Slide Type Pre-cleaned glass slides with smooth ground edges
View More Specs

Products 2
Catalog Number Mfr. No. Mounting Type Price Quantity  
Catalog Number Mfr. No. Mounting Type Price Quantity  
S06041PS View Documents GSC Go Science Crazy
S06041PS
Longitudinal section Each for $6.28
Pack of 10 Each for $62.80
 
S06042PS View Documents GSC Go Science Crazy
S06042PS
Cross section Each for $6.28
Pack of 10 Each for $62.80
 
Description

Description

  • Bundled together in cords and connect to bones via tendons
  • Responsible for movement of animal's skeletal structure
Specifications

Specifications

Prepared Microscope Slides
Zoology
Pre-cleaned glass slides with smooth ground edges
Frog
7 to College