missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Fern Prothallium Microscope Slide

Shows germinating spores filamentous

$7.35 - $7.35

Specifications

For Use With (Application) Shows germinating spores filamentous
Slide Type Pre-cleaned glass slides with smooth ground edges
Species Fern Prothallium
Classification Botany
Type Prepared Microscope Slides
View More Specs

Products 1
Catalog Number Mfr. No. Mounting Type Price Quantity  
Catalog Number Mfr. No. Mounting Type Price Quantity  
S99265PS View Documents GSC Go Science Crazy
S99265PS
Whole mount Each for $7.35
Pack of 10 Each for $73.50
 
Description

Description

  • Prepared slide for use with classroom science studies
Specifications

Specifications

Shows germinating spores filamentous
Fern Prothallium
Prepared Microscope Slides
Pre-cleaned glass slides with smooth ground edges
Botany
7 to College