missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GSC Go Science Crazy Eudiometers

Measures change in volume of gas mixture following a physical or chemical change.

Manufacturer:  GSC Go Science Crazy GIG5032

 View more versions of this product

Catalog No. S32356



Description

Description

  • Capacity: 50 to 100mL
  • Subdivisions at 0.1 to 0.2mL

Specifications

Specifications

Measure the volume of gas produced or consumed in a chemical reaction
Two platinum electrodes
Eudiometer
100mL with subdivisions at 0.2mL
Documents