Please login to your online account to display your discounted pricing

GSC International Eudiometers


Measures change in volume of gas mixture following a physical or chemical change.

Manufacturer: fisher science education  GIG5031

 View more versions of this product

Catalog No. S32353

  • / Each
 

Description & Specifications

Specifications

Class Size 50mL with subdivisions at 0.1mL
For Use With (Application) Measure the volume of gas produced or consumed in a chemical reaction
Includes Two platinum electrodes
Kit Size 50mL with subdivisions at 0.1mL
For Use With (Application) Measure the volume of gas produced or consumed in a chemical reaction
Product Type Eudiometer