Please login to your online account to display your discounted pricing

GSC International Allium Root Microscope Slide

Shows root and root tip

$6.75 - $7.45

Specifications

Type Prepared Microscope Slides
Slide Type Prepared Microscope Slides
Species Allium
Classification Botany
Grade 7 to College
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Price Quantity    

S99211APS

 
gsc international
S99211APS
Shows root tip Each for $7.45
Pack of 10 Each for $74.25

S99270PS

 
gsc international
S99270PS
Shows root Each for $6.75
Pack of 10 Each for $67.50
Description & Specifications

Specifications

Type Prepared Microscope Slides
Slide Type Prepared Microscope Slides
Species Allium
Classification Botany
Grade 7 to College