Please login to your online account to display your discounted pricing

GSC International Allium Root Microscope Slide


Shows root and root tip

Manufacturer: gsc international  S99270PS

 View more versions of this product

Catalog No. S99270PS

  • / Each
  • / Pack of 10 Each
 

Description & Specifications

Specifications

Species Allium
Classification Botany
Grade 7 to College
For Use With (Application) Shows root
Type Prepared Microscope Slides
Slide Type Pre-cleaned glass slides with smooth ground edges