Please login to your online account to display your discounted pricing

GSA 10, Tocris™

Smo receptor agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material GSA 10
Assay Percent Range >98%
Molecular Formula C26H30N2O5
Formula Weight 450.53
Synonym 4-[[(1-Hexyl-1, 2-dihydro-2-oxo-3-quinolinyl)carbonyl]amino]benzoic acid propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

491850

 
r & d systems
RDS491850
50mg Each for $976.15

491810

 
r & d systems
RDS491810
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 450.53
Quantity 50mg
Chemical Name or Material GSA 10
Recommended Storage Store at +4°C
Molecular Formula C26H30N2O5
CAS 300833-95-8
Assay Percent Range >98%
Synonym 4-[[(1-Hexyl-1, 2-dihydro-2-oxo-3-quinolinyl)carbonyl]amino]benzoic acid propyl ester
Safety and Handling

Store at +4°C