missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GS 39783, Tocris

Positive modulator at GABAB receptors

$187.24 - $789.24

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C15H23N5O2S
Formula Weight 337.44
Synonym N, N'-Dicyclopentyl-2-(methylthio)-5-nitro-4, 6-pyrimidinediamine
Chemical Name or Material GS 39783
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
200110 R & D Systems
200110
10mg Each for $187.24
200150 R & D Systems
200150
50mg Each for $789.24
Specifications Description & Specifications

Chemical Identifiers

>98%
337.44
GS 39783
C15H23N5O2S
N, N'-Dicyclopentyl-2-(methylthio)-5-nitro-4, 6-pyrimidinediamine

Specifications

10mg
>98%
337.44
GS 39783
39069-52-8
C15H23N5O2S
N, N'-Dicyclopentyl-2-(methylthio)-5-nitro-4, 6-pyrimidinediamine

Safety and Handling

Recommended Storage : Store at Room Temperature