missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Growth Hormone, Human/hGH Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1328P1ABX

 View more versions of this product

Catalog No. MS1328P1ABX


 

Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Growth Hormone, Human/hGH Ab-3, Mouse Monoclonal Antibody.

hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.

Host Species: Mouse

Clone: GH01/#2 + GH02/#7

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human growth hormone

Molecular Weight: ∽22 kDa

Positive Control: Recombinant human growth hormone, Human anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Growth Hormone, Human/hGH Ab-3
Monoclonal
1mg/mL
Human
Ab without BSA/Azide (Frozen)
Purified recombinant human growth hormone
100μL
Steroids Hormones and Growth Factors
Monoclonal
Immunohistochemistry (Paraffin),Western Blotting
GH01 / #2 + GH02 / #7
Unlabeled
Concentrated
Murine
IgG2a kappa
RUO
Primary
Human