Invitrogen™ GRK7 Recombinant Human Protein DFS Item
GRK7 is a serine/threonine kinase that is specifically expressed in the retina.
Manufacturer:  Invitrogen™ LSPV4125
 View more versions of this product
Catalog No. PV4125
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
GRK7 phosphorylates cone opsins and initiates their deactivation.
Specifications
Specifications
NP_631948
Frozen
89.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
GRK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
GRK7, GPRK7
Baculovirus
Human
GST Tag
Documents