GRK6 Recombinant Human Protein DFS Item
GRK6 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3661
Catalog No. PV3661
Description
Description
GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine⁄threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation.
Specifications
Specifications
NP_001004106
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
94kDa
90% by SDS-PAGE
–80°C
1 Each
GRK6, GPRK6
GRK6 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 5mM MgCl2, 0.25mM EGTA, 0.1mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 240μg/mL partially dephosphorylated casein substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
GRK
29.9nmoles of phosphate transferred to partially dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
0.51mg/mL
2870, 3553
GRK6 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
GRK6 Recombinant Human Protein
Kinase assay
Baculovirus
GRK6 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.