missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GRK6, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002870B01P
Description
Sequence: MELENIVANTVLLKAREGGGGNRKGKSKKWRQMLQFPHISQCEELRLSLERDYHSLCERQPIGRLLFREFCATRPELSRCVAFLDGVAEYEVTPDDKRKACGRQLTQNFLSHTGPDLIPEVPRQLVTNCTQRLEQGPCKDLFQELTRLTHEYLSVAPFADYLDSIYFNRFLQWKWLERQPVTKNTFRQYRVLGKGGFGEVCACQVRATGKMYACKKLEKKRIKKRKGEAMALNEKQILEKVNSRFVVSLAYAYETKDALCLVLTLMNGGDLKFHIYHMGQAGFPEARAVFYAAEICCGLEDLHRERIVYRDLKPENILLDDHGHIRISDLGLAVHVPEGQTIKGRVGTVGYMAPEVVKNERYTFSPDWWALGCFLYEMIAGQSPFQQRKKKIKREEVERLVKEVPEEYSERFSPQARSLCSQLLCKDPAERLGCRGGSAREVKEHPLFKKLNFKRLGAGMLEPPFKPDPQAIYCKDVLDIEQFSTVKGVELEPTDQDFYQKFATGSVPIPWQNEMVETECFQELNVFGLDGSVPPDLDWKGQPPAPPKKGLLQRLFSRQRIAVETAATARKSSPPASSPQPEAPTSSWRSpecifications
GRK6 | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
BC017272.1 | |
GRK6 | |
GRK6 (AAH17272.1, 1 a.a. ∼ 589 a.a) full-length human protein. | |
RUO | |
2870 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
G protein-coupled receptor kinase 6 | |
FLJ32135/GPRK6 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |