Invitrogen™ GRK5 Recombinant Human Protein DFS Item
GRK5 is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4000
 View more versions of this product
Catalog No. PV4000
Description
Description
GRK5 regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes.
Specifications
Specifications
NP_005299
Frozen
95.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
GRK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
GRK5, GPRK5
Baculovirus
Human
GST Tag
Documents