GRK5 Recombinant Human Protein DFS Item
GRK5 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3824
 View more versions of this product
Catalog No. PV3824
Description
Description
GRK5 is a serine⁄threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes.
Specifications
Specifications
NP_005299
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
95.2kDa
>85% by SDS-PAGE
–80°C
1 Each
GRK5, GPRK5
GRK5 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 10μM ATP, 145μg/mL PLKtide peptide substrate (EVIEASFAEQEAK) and trace [32P]- γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
GRK
0.8nmoles of phosphate transferred to PLKtide peptide substrate (EVIEASFAEQEAK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL
GST Tag
0.26mg/mL
2869, 6775
GRK5 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
GRK5 Recombinant Human Protein
Kinase assay
Baculovirus
GRK5 Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.