missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ GRK4 Recombinant Human Protein
GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine/threonine protein kinase family.
Supplier: Invitrogen™ PV3999
Description
GRK4 phosphorylates the activated forms of G protein-coupled receptors.
Specifications
NP_892027 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
GRK4, GPRK4 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
94kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
GRK Kinase Family | |
Recombinant | |
Baculovirus |