Please login to your online account to display your discounted pricing

Thermo Scientific™ GRIP1 (Glucocorticoid Receptor Interacting Protein 1), Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

$279.45 - $1,307.59

Specifications

Antigen GRIP1 (Glucocorticoid Receptor Interacting Protein 1)
Clone GRP01
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1140P

 
thermo scientific™
MS1140P
200μg/mL 1mL Each for $1,307.59

MS1140P0

 
thermo scientific™
MS1140P0
200μg/mL 100µL Each for $279.45

MS1140P1

 
thermo scientific™
MS1140P1
200μg/mL 500µL Each for $822.29
Description & Specifications

Specifications

Antigen GRIP1 (Glucocorticoid Receptor Interacting Protein 1)
Clone GRP01
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Immunogen Recombinant human GRIP1 protein
Isotype IgG2a kappa

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ GRIP1 (Glucocorticoid Receptor Interacting Protein 1), Mouse Monoclonal Antibody.

The steroid hormones, thyroid hormone, vitamin D, and retinoic acid function by activating specific“nuclear receptor proteins” found inside of target cells. The activated receptors bind to and activate specific genes, and the products of these activated genes change cellular activities in specific ways, thus constituting the biological response to the hormone. After binding to target genes, the activated nuclear receptors require the assistance of proteins called“transcriptional co-activators” in order to activate the target genes. GRIP1 (Glucocorticoid Receptor Interacting Protein 1) is a novel protein that serves as a transcriptional co-activator for steroid and nuclear hormone receptors.

Host Species: Mouse

Clone: GRP01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human GRIP1 protein

Molecular Weight: ∽160 kDa

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation
  • Gel Supershift (Not verified)

Note:

USA: RUO; Int'l: RUO