Grey Perch, Bio-Fresh™
Pomadayses magranthus.
$80.00 - $388.73
Specifications
Preservative Bio-Fresh™
View More Specs
Products 2
Catalog Number Mfr. No. Injection Type Length (English) Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Injection Type Length (English) Product Type Quantity Price Quantity  
S9002100 View Documents Fisher Science Education
1341005100
Plain 7 to 9 in. Gray Perch 100/Pail Each for $388.73
S9004 View Documents Fisher Science Education
134098410
Plain 9 to 12 in. Gray Perch 50/CS Pack of 10 Each for $80.00
Case of 5 Pack for $400.00
Description
Description
  • The bony fish specimen of choice in many laboratories has been the yellow perch. In recent years, however, the Mexican grey perch has increasingly been used as a substitute because it can be obtained at a lower cost, yet it provides striking physiological similarities to the yellow perch.
  • We offer specimens in Fisher Science Education™ Bio-Fresh™ or our 100% formaldehyde-free Fisher-Free™ preparation, depending on your needs and preferences
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Specifications
Bio-Fresh™