missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Grey Perch, Bio-Fresh™

Pomadayses magranthus.

Manufacturer:  Fisher Science Education 134098410

 View more versions of this product

Catalog No. S9004




Description

Description

  • The bony fish specimen of choice in many laboratories has been the yellow perch. In recent years, however, the Mexican grey perch has increasingly been used as a substitute because it can be obtained at a lower cost, yet it provides striking physiological similarities to the yellow perch.
  • We offer specimens in Fisher Science Education™ Bio-Fresh™ or our 100% formaldehyde-free Fisher-Free™ preparation, depending on your needs and preferences

Order Info

INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow

Specifications

Specifications

Plain
22.86 to 30.48cm
Bio-Fresh™
50/CS
9 to 12 in.
Vacuum pack
Gray Perch
SDS
Documents
Product Certifications