Please login to your online account to display your discounted pricing

Grey Perch, Bio-Fresh™


Pomadayses magranthus.

Manufacturer: fisher science education  134100510

 View more versions of this product

Catalog No. S900210

  • / Each
 

Description & Specifications

Specifications

Length (English) 7 to 9 in.
Product Type Gray Perch
Preservative Bio-Fresh™
Packaging Pail
Injection Type Plain
Length (Metric) 17.78 to 22.86cm
Quantity 10/Pail