missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One Winged Safety Blood Collection Sets with Holdex™

Pre-attached set with Holdex is a practical, ready-to-use solution

$91.13 - $91.13

Specifications

Includes Holdex Holder
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.
Quantity 24 /pk.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Gauge Price Quantity  
Catalog Number Mfr. No. Gauge Price Quantity  
22-040-165 Greiner Bio-One
450097
21 Pack of 24 Each for $91.13
Case of 10 Pack for $911.30
Specifications

Description

  • Sets are specially developed for blood collection from patients with difficult vein conditions
  • Winged blood collection needle with safety shield provides the necessary protection from the risk of needlestick injury
  • Optical control of blood flow
  • Off-center adapter — ideal angle for venipuncture
  • Can be used with Luer needles
  • Winged needle does not require an adapter
  • Can be connected directly to systems such as indwelling venous catheters
  • Needle length: 0.75 in.

Description & Specifications

Specifications

Holdex Holder
0.75 in.
12 in.
24 /pk.