Please login to your online account to display your discounted pricing
Greiner Bio-One K2EDTA GelTubes
For plasma preparation in molecular diagnostics
$131.83 - $152.63
Specifications
Closure Type Non-ridged pull cap
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
22-040-144
greiner bio-one
454059
Lavender Pack of 50 Each for $131.83
Case of 24 Pack for $3,163.92
22-040-145
greiner bio-one
456058
White Pack of 50 Each for $131.83
Case of 24 Pack for $3,163.92
22-040-67
greiner bio-one
455040
Lavender Pack of 50 Each for $152.63
Case of 24 Pack for $3,663.12
Description & Specifications
Specifications
Closure Type Non-ridged pull cap
  • For detection of viruses, parasites and bacteria
  • interior is spray dried with 1.8mg of anhydrous EDTA per 1mL of blood when tube is appropriately filled
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes should be centrifuged within six hours following blood collection
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.