Greenhouse
Automatic watering makes starting seeds and bulbs easy.
$110.00 - $138.20
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S17622 Fisher Science Education
GRS139
Each for $110.00
S17624 Fisher Scientific
GRS130
Each for $138.20
Specifications
Description
This automatic wick-fed watering system ensures proper moisture level and is a great way to display African violets, orchids, and other exotic plants.
  • Fiberglass wicks never rot
  • Folds up for easy storage
  • Includes the wicks and a 1L reservoir
  • Dimensions: 25H × 22W × 17D in. (62.5 × 55 × 42.5cm)
Order Info
Fluorescent light unit is double tube; fits in greenhouse or terraria (S18450 Series)