Please login to your online account to display your discounted pricing

LaMotte™ GREEN Water Monitoring Kit


An inquiry-based approach to watershed education.

Manufacturer: lamotte™  5848DG

 View more versions of this product

Catalog No. S65402DG

  • / Each
 

Description & Specifications

Specifications

Includes Biochemical oxygen demand, coliform bacteria, dissolved oxygen, nitrate, pH, phosphate, turbidity, temperature, diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous testabs™ reagents
Grade 4+
For Use With (Application) Environmental education
Product Type Green Standard Water Monitoring Kit