Thermo Scientific™ Green Renilla Luciferase Reporter Assay Vectors
Drive Green Renilla luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).
$162.59 - $165.58
Specifications
Quantity 10µg
Formulation DNA at 0.5ug/uL in 10mM Tris-HCl, pH 8.5
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
PI16152 Thermo Scientific™
16152*
pMCS-Green Renilla Luc Vector Each for $162.59
PI16153 Thermo Scientific™
16153*
pCMV-Green Renilla Luc Vector Each for $165.58
PI16154 Thermo Scientific™
16154*
pTK-Green Renilla Luc Vector Each for $163.09
Specifications
Description
The Thermo Scientific™ pMCS-Renilla Luc Vector is a multiple cloning site plasmid to designed to accept a promoter sequence for study of gene regulation using the intracellular Renilla luciferase reporter.
The Green Renilla Luc Vectors contain a gene cloned from the sea pansy, Renilla reniformis. The gene encodes a bioluminescent Green Renilla luciferase (36kDa). The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.
Highlights:
  • Intracellular Green Renilla luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
Description & Specifications
Specifications
10µg
DNA at 0.5ug/uL in 10mM Tris-HCl, pH 8.5