missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite tape, square weave, 0.56mm (0.022 in.) thick x 100mm (3.94in) wide, 99.9%

Manufacturer:  Alfa Aesar™ 10136BU

 View more versions of this product

Catalog No. AA10136BU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Graphite tape
Square weave, 0.56mm (0.022 in.) thick x 100mm (3.94 in.) wide
99.9%
25cm
MFCD00144065

Chemical Identifiers

MFCD00144065
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents